Learn More
Bochem™ Zinced Pinchcock with Aluminum Screw
Restrict or regulate tubing fluid flow with these zinced, blade-style pinchcocks. Bochem™ Zinced Pinchcocks facilitates precise flow adjustment.
Brand: Bochem™ 12121
Additional Details : Weight : 0.80000kg
Description
- Precisely and easily adjust flow.
- Durable and resistant.
Specifications
Pinchcock | |
15 mm | |
Zinc, Aluminum |