missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Zinced Pinchcock with Aluminum Screw
Restrict or regulate tubing fluid flow with these zinced, blade-style pinchcocks. Bochem™ Zinced Pinchcocks facilitates precise flow adjustment.
16.35 EUR - 17.80 EUR
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10182501
|
Bochem™
12121 |
15 mm |
16.35 EUR
Each |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||||
10428432
|
Bochem™
12122 |
20 mm |
17.80 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
Description
- Precisely and easily adjust flow.
- Durable and resistant.
Specifications
Pinchcock | |
Zinc, Aluminum |