Learn More
1,3,5-Trifluorobenzene, 98+%, Thermo Scientific Chemicals
74.67 EUR - 1002.99 EUR
Chemical Identifiers
CAS | 372-38-3 |
---|---|
Molecular Formula | C6H3F3 |
Molecular Weight (g/mol) | 132.085 |
MDL Number | MFCD00000333 |
InChI Key | JXUKFFRPLNTYIV-UHFFFAOYSA-N |
Synonym | benzene, 1,3,5-trifluoro, sym-trifluorobenzene, unii-bn94c411f8, pubchem1069, 1,5-trifluorobenzene, acmc-209iru, 2,4,6-trifluorophenyl, benzene,3,5-trifluoro, 1,3,5-tri-fluorobenzene, 1,3,5-trifluoro-benzene |
PubChem CID | 9745 |
IUPAC Name | 1,3,5-trifluorobenzene |
SMILES | C1=C(C=C(C=C1F)F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11458566
|
Thermo Scientific Alfa Aesar
A15984.06 |
5 g |
74.67 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11468566
|
Thermo Scientific Alfa Aesar
A15984.14 |
25 g |
249.24 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11478566
|
Thermo Scientific Alfa Aesar
A15984.22 |
100 g |
1002.99 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,3,5-Trifluorobenzene was used in the synthesis of (-)-epicatechin and its 3-O-gallate. It is also used as an intermediate in organic chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3,5-Trifluorobenzene was used in the synthesis of (-)-epicatechin and its 3-O-gallate. It is also used as an intermediate in organic chemical synthesis.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
372-38-3 | |
132.085 | |
JXUKFFRPLNTYIV-UHFFFAOYSA-N | |
9745 | |
C1=C(C=C(C=C1F)F)F |
C6H3F3 | |
MFCD00000333 | |
benzene, 1,3,5-trifluoro, sym-trifluorobenzene, unii-bn94c411f8, pubchem1069, 1,5-trifluorobenzene, acmc-209iru, 2,4,6-trifluorophenyl, benzene,3,5-trifluoro, 1,3,5-tri-fluorobenzene, 1,3,5-trifluoro-benzene | |
1,3,5-trifluorobenzene |
Specifications
372-38-3 | |
1.279 | |
−7°C (19°F) | |
1.415 | |
UN2387 | |
benzene, 1,3,5-trifluoro, sym-trifluorobenzene, unii-bn94c411f8, pubchem1069, 1,5-trifluorobenzene, acmc-209iru, 2,4,6-trifluorophenyl, benzene,3,5-trifluoro, 1,3,5-tri-fluorobenzene, 1,3,5-trifluoro-benzene | |
JXUKFFRPLNTYIV-UHFFFAOYSA-N | |
1,3,5-trifluorobenzene | |
9745 | |
≥98% |
-7°C to -4°C | |
75°C to 76°C | |
C6H3F3 | |
MFCD00000333 | |
1906457 | |
Insoluble in water. | |
C1=C(C=C(C=C1F)F)F | |
132.085 | |
132.09 | |
1,3,5-Trifluorobenzene |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROBENZENE
missing translation for 'einecsNumber' : 206-751-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.