missing translation for 'onlineSavingsMsg'
Learn More

1,4-Phenylenediacetonitrile, 97%, Thermo Scientific Chemicals

42.01 EUR - 144.72 EUR

Chemical Identifiers

CAS 622-75-3
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.188
MDL Number MFCD00001923
InChI Key FUQCKESKNZBNOG-UHFFFAOYSA-N
Synonym 1,4-phenylenediacetonitrile, 1,4-benzenediacetonitrile, p-xylylene dicyanide, p-phenylenediacetonitrile, p-benzenediacetonitrile, 2,2'-1,4-phenylene diacetonitrile, p-bis cyanomethyl benzene, 1,4-bis cyanomethyl benzene, p-cyanomethyl benzyl cyanide, 2-4-cyanomethyl phenyl acetonitrile
PubChem CID 69327
IUPAC Name 2-[4-(cyanomethyl)phenyl]acetonitrile
SMILES C1=CC(=CC=C1CC#N)CC#N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11353087
SDS View Documents
Thermo Scientific Alfa Aesar
L13464.06
5 g
42.01 EUR
5g
Please to purchase this item. Need a web account? Register with us today!
11363087
SDS View Documents
Thermo Scientific Alfa Aesar
L13464.14
25 g
144.72 EUR
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1,4-Phenylenediacetonitrile, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Phenylenediacetonitrile, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

missing translation for 'einecsNumber' : 210-751-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,4-Phenylenediacetonitrile, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.