Learn More
1S,3R-RSL3
Induces ferroptosis; inhibits glutathione peroxidase 4 (GPX4)
Brand:  Tocris Bioscience 6118/50
 View more versions of this product
Product Code. 18749278
  • 1210.00 EUR / 50mg
Explore more special offers
Description
Description
1S,3R-RSL3 is a glutathione peroxidase 4 (GPX4) inhibitor. Induces ferroptosis. Increases lipid peroxidase levels in vitro. Inhibits tumor formation and progression in vivo.
Negative control 1R,3R-RSL3 (Cat. No. 6687) also available.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
(1S,3R)-Methyl 2-(2-chloroacetyl)-2,3,4,9-tetrahydro-1-[4-(methoxycarbonyl)phenyl]-1H-pyrido[3,4-b]indole-3-carboxylate
>98%
C23H21ClN2O5
1219810-16-8
50 mg
Observed MW: 440.88
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
1S,3R-RSL3 > 50mg