missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1S,3R-RSL3
Induces ferroptosis; inhibits glutathione peroxidase 4 (GPX4)
313.00 EUR - 1210.00 EUR
Specifications
Chemical Name or Material | (1S,3R)-Methyl 2-(2-chloroacetyl)-2,3,4,9-tetrahydro-1-[4-(methoxycarbonyl)phenyl]-1H-pyrido[3,4-b]indole-3-carboxylate |
---|---|
CAS | 1219810-16-8 |
Assay Percent Range | >98% |
Molecular Formula | C23H21ClN2O5 |
Formula Weight | Observed MW: 440.88 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18773999
|
Tocris Bioscience
6118/10 |
10 mg |
313.00 EUR
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18749278
|
Tocris Bioscience
6118/50 |
50 mg |
1210.00 EUR
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glutathione peroxidase 4 (GPX4) inhibitor. Induces ferroptosis. Increases lipid peroxidase levels in vitro. Inhibits tumor formation and progression in vivo.Specifications
(1S,3R)-Methyl 2-(2-chloroacetyl)-2,3,4,9-tetrahydro-1-[4-(methoxycarbonyl)phenyl]-1H-pyrido[3,4-b]indole-3-carboxylate | |
>98% | |
Observed MW: 440.88 |
1219810-16-8 | |
C23H21ClN2O5 |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1S,3R-RSL3