Learn More
2,3-O-Isopropylidene-alpha,beta-D-ribofuranose, Thermo Scientific Chemicals
70.85 EUR - 253.26 EUR
Chemical Identifiers
CAS | 13199-25-2 |
---|---|
Molecular Formula | C8H14O5 |
MDL Number | MFCD01075718 |
Description
2,3-O-Isopropylidene-alpha,beta-D-ribofuranose is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,3-O-Isopropylidene-alpha,beta-D-ribofuranose is used as a pharmaceutical intermediate.
Solubility
Soluble in acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
13199-25-2 | |
C8H14O5 | |
MFCD01075718 |
Specifications
13199-25-2 | |
MFCD01075718 | |
−27° (c=1 in Chloroform) | |
2,3-O-Isopropylidene-alpha,beta-D-ribofuranose |
C8H14O5 | |
Soluble in acetone. | |
190.2 |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.