missing translation for 'onlineSavingsMsg'
Learn More
2,4-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals Product Code.: 11475293

2,4-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 11475293
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11475293

Brand: Thermo Scientific Alfa Aesar A12868.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2,4-Dihydroxybenzaldehyde is used in two-step synthesis of ethyl 3,5-dibromo-2,4-dihydroxycinnamate. It has a potent antioxidative and antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dihydroxybenzaldehyde is used in two-step synthesis of ethyl 3,5-dibromo-2,4-dihydroxycinnamate. It has a potent antioxidative and antibacterial activity.

Solubility
Soluble in water. Solubility in methanol is almost transparent. Easily soluble in alcohol, ethyl ether, chloroform, glacial acetic acid. Insoluble in benzene and easily decomposes under strong acid strong alkali.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-01-2
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00011686
InChI Key IUNJCFABHJZSKB-UHFFFAOYSA-N
Synonym beta-resorcylaldehyde, 4-hydroxysalicylaldehyde, 4-formylresorcinol, beta-resorcaldehyde, benzaldehyde, 2,4-dihydroxy, beta-resorcinaldehyde, beta-resorcylic aldehyde, 2,4-dihydroxybenzenecarbonal, salicylaldehyde, 4-hydroxy, 4-hydroxysalicyladehydeShow MoreShow Less
PubChem CID 7213
ChEBI CHEBI:50198
IUPAC Name 2,4-dihydroxybenzaldehyde
SMILES OC1=CC=C(C=O)C(O)=C1

Specifications

Melting Point 134°C to 138°C
Boiling Point 220°C (22 mmHg)
Quantity 100 g
Beilstein 878548
Sensitivity Air Sensitive
Merck Index 14,8156
Solubility Information Soluble in water. Solubility in methanol is almost transparent. Easily soluble in alcohol,ethyl ether,chloroform,glacial acetic acid. Insoluble in benzene and easily decomposes under strong acid strong alkali.
Formula Weight 138.12
Percent Purity 98%
Chemical Name or Material 2,4-Dihydroxybenzaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10236172 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA12868.22,A12868.14,A12868.36
Lot Number10249719 Certificate TypeCertificate of Analysis Date22/02/2024 Catalog NumberA12868.22,A12868.14,A12868.36
Lot Number10154070 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12868.22,A12868.14,A12868.36
Lot Number10182188 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12868.22,A12868.14,A12868.36
Lot Number10161971 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12868.22,A12868.14,A12868.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.