missing translation for 'onlineSavingsMsg'
Learn More
2,4-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals Product Code.: 10479310

2,4-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 10479310
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10479310

Brand: Thermo Scientific Acros 173641000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-01-2
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00011686
InChI Key IUNJCFABHJZSKB-UHFFFAOYSA-N
Synonym beta-resorcylaldehyde, 4-hydroxysalicylaldehyde, 4-formylresorcinol, beta-resorcaldehyde, benzaldehyde, 2,4-dihydroxy, beta-resorcinaldehyde, beta-resorcylic aldehyde, 2,4-dihydroxybenzenecarbonal, salicylaldehyde, 4-hydroxy, 4-hydroxysalicyladehydeShow MoreShow Less
PubChem CID 7213
ChEBI CHEBI:50198
IUPAC Name 2,4-dihydroxybenzaldehyde
SMILES OC1=CC=C(C=O)C(O)=C1

Specifications

Melting Point 133.0°C to 138.0°C
Color White to Beige
Boiling Point 220.0°C to 228.0°C (22.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (GC)
Packaging Glass bottle
Linear Formula (HO)2C6H3CHO
Quantity 100 g
Beilstein 08,241
Merck Index 15,8277
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in alcohol,ether,chloroform and,glacial acetic acic
Formula Weight 138.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Dihydroxybenzaldehyde
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0459543 Certificate TypeCertificate of Analysis Date29/01/2024 Catalog Number173645000,173640250,173641000
Lot NumberA0456138 Certificate TypeCertificate of Analysis Date06/09/2023 Catalog Number173645000,173640250,173641000
Lot NumberA0453843 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number173645000,173640250,173641000
Lot NumberA0431988 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog Number173645000,173640250,173641000
Lot NumberA0399963 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number173645000,173640250,173641000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.