Learn More
4-(Trifluoroacetyl)toluene, 97%, Thermo Scientific Chemicals
71.15 EUR - 261.30 EUR
Chemical Identifiers
CAS | 394-59-2 |
---|---|
Molecular Formula | C9H7F3O |
Molecular Weight (g/mol) | 188.149 |
MDL Number | MFCD00052338 |
InChI Key | DYILUJUELMWXAL-UHFFFAOYSA-N |
Synonym | 4-trifluoroacetyl toluene, 2,2,2-trifluoro-1-p-tolyl ethanone, 4'-methyl-2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-p-tolylethanone, 2,2,2-trifluoro-1-4-methylphenyl ethanone, 2,2,2-trifluoro-4'-methylacetophenone, 2,2,2-trifluoro-1-4-methylphenyl ethan-1-one, 2,2,2-trifluoro-1-p-tolyl-ethanone, 4-trifluoroacetyltoluene, acmc-209j5mShow More |
PubChem CID | 136237 |
IUPAC Name | 2,2,2-trifluoro-1-(4-methylphenyl)ethanone |
SMILES | CC1=CC=C(C=C1)C(=O)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11378795
|
Thermo Scientific Alfa Aesar
L12424.03 |
1 g |
71.15 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11388795
|
Thermo Scientific Alfa Aesar
L12424.06 |
5 g |
261.30 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
394-59-2 | |
188.149 | |
DYILUJUELMWXAL-UHFFFAOYSA-N | |
136237 | |
CC1=CC=C(C=C1)C(=O)C(F)(F)F |
C9H7F3O | |
MFCD00052338 | |
4-trifluoroacetyl toluene, 2,2,2-trifluoro-1-p-tolyl ethanone, 4'-methyl-2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-p-tolylethanone, 2,2,2-trifluoro-1-4-methylphenyl ethanone, 2,2,2-trifluoro-4'-methylacetophenone, 2,2,2-trifluoro-1-4-methylphenyl ethan-1-one, 2,2,2-trifluoro-1-p-tolyl-ethanone, 4-trifluoroacetyltoluene, acmc-209j5mShow More | |
2,2,2-trifluoro-1-(4-methylphenyl)ethanone |
Specifications
394-59-2 | |
C9H7F3O | |
MFCD00052338 | |
4-trifluoroacetyl toluene, 2,2,2-trifluoro-1-p-tolyl ethanone, 4'-methyl-2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-p-tolylethanone, 2,2,2-trifluoro-1-4-methylphenyl ethanone, 2,2,2-trifluoro-4'-methylacetophenone, 2,2,2-trifluoro-1-4-methylphenyl ethan-1-one, 2,2,2-trifluoro-1-p-tolyl-ethanone, 4-trifluoroacetyltoluene, acmc-209j5m | |
CC1=CC=C(C=C1)C(=O)C(F)(F)F | |
188.149 | |
188.15 | |
4-(Trifluoroacetyl)toluene |
70°C to 72°C (15 mmHg) | |
1.47 | |
1870406 | |
DYILUJUELMWXAL-UHFFFAOYSA-N | |
2,2,2-trifluoro-1-(4-methylphenyl)ethanone | |
136237 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.