Learn More
Hamilton™ 50 μL 1800 Series Gas Tight Syringe
The 1800 Series Gas Tight syringes are the reinforced plunger version of the 1700 Series.
Brand:  Hamilton™ CAL7638-01
 View more versions of this product
Product Code. 17854232
  • 331.00 EUR / Each
Explore more special offers
Description
Description
  • Attached to the flange of the syringe is a blue syringe holder, which provides an area to hold the syringe that prevents heat transfer, dispense inaccuracies, and plunger blow outs.
  • Provides additional support to small-volume plungers that are prone to bending during injection.
  • This series of syringes are ideal for dispensing volumes up to 50 μL.
Specifications
Specifications
Not Autoclavable
22s G
0.0059 in.
0.0283 in.
0.729 mm
0.72 mm
122°F
50°F
10°C
50 μL
Sharp, Bevel, Curved, Non-coring
PTFE
0.029 in.
0.15 mm
0.305 in.
7.75 mm
2 in.
50°C
1805 N
1
51 mm
Cemented
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hamilton™ 50 μL 1800 Series Gas Tight Syringe > Needle Gauge: 22s G, Point Style: 2, Connection Type: N