Learn More
BINDER™ Access Port
Access port with silicone plug for use with KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, and KBF PRO 1600.
Brand:  BINDER™ 8012-2492
 View more versions of this product
Product Code. 30238212
  • 505.00 EUR / Each
Estimated Shipment: 29-04-2025
Description
  • Round access port with silicone plug
  • Choose between 30, 50, or 100 mm plug
  • Available on left, right, or top of the chamber
  • For use with KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, and KBF PRO 1600
Specifications
Specifications
KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, KBF PRO 1600
Access Port
30 mm
Silicone
1 on Right Side
Product Title
BINDER™ Access Port > Access Port, 30 mm, Right