Learn More
BINDER™ Access Port
Access port with silicone plug for use with KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, and KBF PRO 1600.
505.00 EUR - 943.00 EUR
Specifications
For Use With (Equipment) | KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, KBF PRO 1600 |
---|---|
Material | Silicone |
Type | Access Port |
Product Code | Brand | Port Size | Ports | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Port Size | Ports | Price | Quantity & Availability | |||||
30238386
|
BINDER™
8012-2489 |
30 mm | 1 on Left Side |
505.00 EUR
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
|||||
30238372
|
BINDER™
8012-2490 |
50 mm | 1 on Left Side |
589.00 EUR
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
|||||
30238236
|
BINDER™
8012-2491 |
100 mm | 1 on Left Side |
943.00 EUR
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
|||||
30238212
|
BINDER™
8012-2492 |
30 mm | 1 on Right Side |
505.00 EUR
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
|||||
30238366
|
BINDER™
8012-2493 |
50 mm | 1 on Right Side |
589.00 EUR
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
|||||
30238269
|
BINDER™
8012-2494 |
100 mm | 1 on Right Side |
943.00 EUR
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
|||||
30238341
|
BINDER™
8012-2495 |
50 mm | 1 on Top |
589.00 EUR
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
|||||
30238197
|
BINDER™
8012-2496 |
100 mm | 1 on Top |
943.00 EUR
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
|||||
30238368
|
BINDER™
8012-2500 |
30 mm | 1 on Top |
505.00 EUR
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
|||||
Description
- Round access port with silicone plug
- Choose between 30, 50, or 100 mm plug
- Available on left, right, or top of the chamber
- For use with KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, and KBF PRO 1600
Specifications
KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, KBF PRO 1600 | |
Silicone | |
Access Port |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BINDER™ Access Port