missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BINDER™ Access Port
Access port with silicone plug for use with KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, and KBF 1600.
Brand: BINDER™ 8012-2487
This item is not returnable.
View return policy
Description
- Round access port with silicone plug
- Choose between 30, 50, or 100 mm plug
- Available on left, right, or top of the chamber
- For use with KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, and KBF 1600
Specifications
KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, KBF 1600 | |
Access Port | |
50 mm |
Silicone | |
1 on Top |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BINDER™ Access Port > Access Port, 50 mm, Top
Spot an opportunity for improvement?Share a Content Correction