Learn More
3M™ Adflo™ Powered Air Purifying Respirator System with 3M™ Speedglas™ G5-01 Welding Helmet
3M™ Adflo™ Powered Air Purifying Respirator System with 3M™ Speedglas™ G5-01 Series Welding Helmet provides all of the benefits of our G5-01 series welding helmet with our convenient Adflo™ Powered Air Purifying Respirator system.
Brand: 3M™ 7100257943
1857.56 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For workers in heavy duty environments, 3M™ Adflo™ Powered Air Purifying Respirator with Speedglas™ G5-01 Series Heavy-Duty Welding Helmet system provides High-Efficiency particulate respiratory protection for welders in a robust yet compact design with ergonomic, adjustable head suspension.
- Flip front welding shield allows for a clear, wide-view protective visor for grinding and setup.
- The 3M™ Adflo™ Powered Air Respirator has a High-Efficiency particulate filter that can be stacked onto an optional 3M™ Adflo™ Filter (not included) for additional protection against certain vapours or a Nuisance Odour Pad Assembly.
- Audible and visual alarms signal low airflow and low battery charge.
- Available in two different Auto-Darkenig Lens options (VC/TW). G5 Series storage/carry bag included.
Specifications
7 to 9 hr. | |
COMBINATION | |
G5-01VC | |
One Size | |
No |
Lithium-ion | |
Black | |
PAPR System for Welding | |
PAPR, Welding Helmet |
Product Suggestions
Customers who viewed this item also viewed
CE_17895666_EN.pdf
File size: 926.9 kB
Product Content Correction
Product Title
3M™ Adflo™ Powered Air Purifying Respirator System with 3M™ Speedglas™ G5-01 Welding Helmet > Filter: G5-01VC
Spot an opportunity for improvement?Share a Content Correction