Learn More
3M™ Adflo™ Powered Air Purifying Respirator System with 3M™ Speedglas™ G5-01 Welding Helmet
3M™ Adflo™ Powered Air Purifying Respirator System with 3M™ Speedglas™ G5-01 Series Welding Helmet provides all of the benefits of our G5-01 series welding helmet with our convenient Adflo™ Powered Air Purifying Respirator system.
2462.78 EUR - 2607.72 EUR
Specifications
Cartridge/Filter | COMBINATION |
---|---|
Color | Black |
Product Type | PAPR System for Welding |
Size | One Size |
Includes | PAPR, Welding Helmet |
Product Code | Brand | Filter Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filter Type | Price | Quantity & Availability | ||||||
17885666
|
3M™
7100256131 |
G5-01TW | N/A | |||||||
17895666
|
3M™
7100257943 |
G5-01VC |
2607.72 EUR
Each |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||||
Description
- For workers in heavy duty environments, 3M™ Adflo™ Powered Air Purifying Respirator with Speedglas™ G5-01 Series Heavy-Duty Welding Helmet system provides High-Efficiency particulate respiratory protection for welders in a robust yet compact design with ergonomic, adjustable head suspension.
- Flip front welding shield allows for a clear, wide-view protective visor for grinding and setup.
- The 3M™ Adflo™ Powered Air Respirator has a High-Efficiency particulate filter that can be stacked onto an optional 3M™ Adflo™ Filter (not included) for additional protection against certain vapours or a Nuisance Odour Pad Assembly.
- Audible and visual alarms signal low airflow and low battery charge.
- Available in two different Auto-Darkenig Lens options (VC/TW). G5 Series storage/carry bag included.
Specifications
COMBINATION | |
PAPR System for Welding | |
PAPR, Welding Helmet |
Black | |
One Size | |
No |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
3M™ Adflo™ Powered Air Purifying Respirator System with 3M™ Speedglas™ G5-01 Welding Helmet