missing translation for 'onlineSavingsMsg'
Learn More
Ammonia, 25% w/w, Reagent Grade, Scharlab Product Code.: 11309823
compliance-icons compliance-icons compliance-icons

Ammonia, 25% w/w, Reagent Grade, Scharlab

Product Code. 11309823
1 L
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 11309823

Brand: Scharlab AM02491000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Laboratory reagent for use in analytical chemistry.

  • Synonyms: Ammonia water, Ammonium hydroxide solution
  • Molecular Formula: NH3
  • Molar Mass: 17.03g/mol
  • CAS: 1336-21-6
  • EIN ECS No.: 215-647-6

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 ohShow MoreShow Less
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Boiling Point 37.7°C
Melting Point -57.5°C
Color Colorless
Physical Form Liquid
Quantity 1 L
Formula Weight 17.03g/mol
Percent Purity 25 to 30% (acidimetric, NH3)
Grade Reagent
Vapor Pressure ∼500hPa at 20°C
Density 0.90g/cm3
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonia, solution 25% w/w, reagent grade, Reag. Ph Eur
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Ammonia
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonia, 25% w/w, Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.