Learn More
Oxoid UK Anaerobic 2.5 Litre Rectangular Jar Complete
Anaerobic 2.5 Litre Rectangular Jar Complete Stackable
Brand: Oxoid UK AB0025A
296.58 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Anaerobic 2.5 Litre Rectangular Jar Complete Stackable holds up to 12x 90mm plates or 21 x 55mm plates
Specifications
Rectangular Jar | |
Each |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6206621 | Certificate of Analysis | 29/11/2024 | AB0025A |
6113172 | Certificate of Analysis | 03/05/2024 | AB0025A |
3780085 | Certificate of Analysis | 24/01/2024 | AB0025A |
3614354 | Certificate of Analysis | 04/02/2023 | AB0025A |
3615130 | Certificate of Analysis | 04/02/2023 | AB0025A |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
76Hours
21Minutes
54Seconds
45New Offer Save Up to 85%