Learn More
PCNA Mouse anti-Human, Mouse, Rat, Porcine, Chicken, Drosophila, Fish, Marsupial, Primate, Rabbit, Yeast, Zebrafish, Clone: PC10, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 35 publications
Brand:  Novus Biologicals NB500-106
Encompass
Product Code. 12862006
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
PCNA Monoclonal specifically detects PCNA in Human, Mouse, Rat, Porcine, Chicken, Drosophila, Fish, Marsupial, Primate, Rabbit, Yeast, Zebrafish samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
PCNA Mouse anti-Human, Mouse, Rat, Porcine, Chicken, Drosophila, Fish, Marsupial, Primate, Rabbit, Yeast, Zebrafish, Clone: PC10, Novus Biologicals™