missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNA Antibody (PC10), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 35 publications
Brand: Novus Biologicals NB500-106
This item is not returnable.
View return policy
Description
PCNA Monoclonal specifically detects PCNA in Human, Mouse, Rat, Porcine, Chicken, Drosophila, Fish, Marsupial, Primate, Rabbit, Yeast, Zebrafish samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.
Specifications
PCNA | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Simple Western 1:200, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 26883631), Flow Cytometry 1:10-1:1000, ELISA reported in scientific literature, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:1000-1:2000. Use reported in scientific literature (PMID 22068968), Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen 1:10-1:500, Immunohistochemistry Free-Floating reported in scientific literature (PMID 27212918), Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen | |
Mouse | |
30 kDa | |
0.1 mL | |
Autophagy, Base Excision Repair, Cellular Markers, DNA Repair, Loading Controls | |
5111 | |
Human, Mouse, Rat, Pig, Chicken, Drosophila, Fish, Marsupial, Primate, Rabbit, Yeast, Zebrafish | |
Purified |
Western Blot, ChIP Assay, Flow Cytometry, ELISA, Immunohistochemistry | |
PC10 | |
Unconjugated | |
P12004 | |
PCNA | |
Protein A-rat PCNA fusion obtained from pC2T | |
Protein A purified | |
RUO | |
Primary | |
Specific for PCNA p36 protein expressed at high levels in proliferating cells. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a κ |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PCNA Antibody (PC10), Novus Biologicals™ > 0.1mL; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction