missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNA Antibody (PC10), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 35 publications
223.00 EUR - 562.00 EUR
Specifications
Antigen | PCNA |
---|---|
Clone | PC10 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:2000, Simple Western 1:200, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 26883631), Flow Cytometry 1:10-1:1000, ELISA reported in scientific literature, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:1000-1:2000. Use reported in scientific literature (PMID 22068968), Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen 1:10-1:500, Immunohistochemistry Free-Floating reported in scientific literature (PMID 27212918), Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
Applications | Western Blot, ChIP Assay, Flow Cytometry, ELISA, Immunohistochemistry |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18776643
|
Novus Biologicals
NB500-106SS |
0.025 mL |
223.00 EUR
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18730194
|
Novus Biologicals
NB500-106 |
0.1 mL |
562.00 EUR
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PCNA Monoclonal specifically detects PCNA in Human, Mouse, Rat, Porcine, Chicken, Drosophila, Fish, Marsupial, Primate, Rabbit, Yeast, Zebrafish samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.Specifications
PCNA | |
1 mg/mL | |
Western Blot, ChIP Assay, Flow Cytometry, ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Autophagy, Base Excision Repair, Cellular Markers, DNA Repair, Loading Controls | |
P12004 | |
5111 | |
Protein A-rat PCNA fusion obtained from pC2T | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
30 kDa |
PC10 | |
Western Blot 1:2000, Simple Western 1:200, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 26883631), Flow Cytometry 1:10-1:1000, ELISA reported in scientific literature, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:1000-1:2000. Use reported in scientific literature (PMID 22068968), Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen 1:10-1:500, Immunohistochemistry Free-Floating reported in scientific literature (PMID 27212918), Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Pig, Chicken, Drosophila, Fish, Marsupial, Primate, Rabbit, Yeast, Zebrafish | |
cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen | |
PCNA | |
IgG2a κ | |
Protein A purified | |
Specific for PCNA p36 protein expressed at high levels in proliferating cells. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PCNA Antibody (PC10), Novus Biologicals™