Learn More
Erlab™ Captair Bio 391 Workstation
Erlab™ Captair Bio 391 Workstation features a HEPA H14 filter, stainless steel work surface, and a UV light with a timer that provides the proper sterilization to protect your samples from contamination.
Brand:  Erlab™ E12F/AS
Product Code. 15890295
Description
Description
  • High efficiency particulate filtration
  • Simple, intuitive communication by light shows the status of the hood
  • Internal UV light
  • Air flow monitoring
  • Internal LED lighting
  • UV resistant front and side panels
  • Ergonomic design
  • Easy to clean
  • Mobile or fixed benches
  • Filter HP - HEPA H14 : This filtration technology traps particles larger than 0.1 μm with 99.995
  • Adding a carbon filter to your enclosure allows protection of your samples from VOCs
  • Filter AS for organic vapors
Specifications
Specifications
Mobile Ductless Fiiltering PCR Workstation
630 x 1030 x 945 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Erlab™ Captair Bio 391 Workstation > Dimensions (L x W x H): 630 x 1030 x 945 mm