missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Lambda Fab Select Leakage ELISA

Product Code. 18436508 Shop All Thermo Scientific Products
10 Assays, Pack of 10
Click to view available options
Quantity:
1 Assay
10 Assays
Unit Size:
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 18436508

Brand: Thermo Scientific™ 810308010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ LambdaFabSelect Leakage ELISA is designed to detect possible leached ligand of human Fab fragments or Ig's with a lambda light chain that have been purified using LambdaFabSelect affinity resin (Cytiva).

CaptureSelect LambdaFabSelect Leakage ELISA is designed to detect possible leached ligand of human Fab fragments or Ig's with a lambda light chain that have been purified using LambdaFabSelect affinity resin (Cytiva). Even when covalently attached, small amounts of the LambdaFabSelect affinity ligand can leach off of the chromatography support and co-elute with the antibody.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect LambdaFabSelect Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

Specifications

Content And Storage Store at -5°C to -30°C.
Assay ELISA
For Use With (Equipment) Microplate Reader
Product Line CaptureSelect™
Quantity 10 Assays
Target LambdaFabSelect Affinity Ligand
Type ELISA Kit
Format Kit

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Product Code
Lot Number010616-05 Certificate TypeCertificate of Analysis Date23/10/2020 Product Code810308010
Lot Number010616-04 Certificate TypeCertificate of Analysis Date21/09/2020 Product Code810308010
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ CaptureSelect™ Lambda Fab Select Leakage ELISA >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.