Learn More
Thermo Scientific™ CaptureSelect™ Lambda Fab Select Leakage ELISA
Thermo Scientific™ CaptureSelect™ LambdaFabSelect Leakage ELISA is designed to detect possible leached ligand of human Fab fragments or Ig's with a lambda light chain that have been purified using LambdaFabSelect affinity resin (Cytiva).
1065.00 EUR - 9240.00 EUR
Specifications
Content And Storage | Store at -5°C to -30°C. |
---|---|
Assay | ELISA |
For Use With (Equipment) | Microplate Reader |
Product Line | CaptureSelect™ |
Target | LambdaFabSelect Affinity Ligand |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
18406748
|
Thermo Scientific™
810308001 |
1 Assay |
1065.00 EUR
Each |
In Stock
Log in to see stock. |
||||||
18436508
|
Thermo Scientific™
810308010 |
10 Assays |
9240.00 EUR
Pack of 10 |
In Stock
Log in to see stock. |
||||||
Description
CaptureSelect LambdaFabSelect Leakage ELISA is designed to detect possible leached ligand of human Fab fragments or Ig's with a lambda light chain that have been purified using LambdaFabSelect affinity resin (Cytiva). Even when covalently attached, small amounts of the LambdaFabSelect affinity ligand can leach off of the chromatography support and co-elute with the antibody.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect LambdaFabSelect Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Specifications
Store at -5°C to -30°C. | |
Microplate Reader | |
LambdaFabSelect Affinity Ligand | |
Kit |
ELISA | |
CaptureSelect™ | |
ELISA Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.