missing translation for 'onlineSavingsMsg'
Learn More

Chrome Azurol S, MP Biomedicals™

Chrome Azurol S is used as a reagent for determination of fluoride

45.05 EUR - 314.00 EUR

Chemical Identifiers

CAS 1667-99-8
Molecular Formula C23H13Cl2Na3O9S
Molecular Weight (g/mol) 605.277
MDL Number MFCD00001615
InChI Key FUIZKNBTOOKONL-UHFFFAOYSA-K
Synonym Mordant Blue 29, C.I. 43825
PubChem CID 54736314
IUPAC Name trisodium;5-[(3-carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)-(2,6-dichloro-3-sulfonatophenyl)methyl]-3-methyl-2-oxidobenzoate
SMILES CC1=CC(=CC(=C1[O-])C(=O)[O-])C(=C2C=C(C(=O)C(=C2)C(=O)O)C)C3=C(C=CC(=C3Cl)S(=O)(=O)[O-])Cl.[Na+].[Na+].[Na+]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11493182
View Documents
MP Biomedicals
0215498210
10 g
45.05 EUR
10g
Estimated Shipment: 19-03-2025
to see stock.
Add to basket
 
11427554
View Documents
MP Biomedicals
0520265510
10 g
54.20 EUR
10g
Estimated Shipment: 19-03-2025
to see stock.
Add to basket
 
11403192
View Documents
MP Biomedicals
0215498225
25 g
86.75 EUR
25g
Estimated Shipment: 19-03-2025
to see stock.
Add to basket
 
11413192
View Documents
MP Biomedicals
0215498280
100 g
314.00 EUR
100g
Estimated Shipment: 19-03-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Chemical Identifiers

1667-99-8
605.277
FUIZKNBTOOKONL-UHFFFAOYSA-K
54736314
CC1=CC(=CC(=C1[O-])C(=O)[O-])C(=C2C=C(C(=O)C(=C2)C(=O)O)C)C3=C(C=CC(=C3Cl)S(=O)(=O)[O-])Cl.[Na+].[Na+].[Na+]
C23H13Cl2Na3O9S
MFCD00001615
Mordant Blue 29, C.I. 43825
trisodium;5-[(3-carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)-(2,6-dichloro-3-sulfonatophenyl)methyl]-3-methyl-2-oxidobenzoate

Specifications

1667-99-8
MFCD00001615
FUIZKNBTOOKONL-UHFFFAOYSA-K
trisodium;5-[(3-carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)-(2,6-dichloro-3-sulfonatophenyl)methyl]-3-methyl-2-oxidobenzoate
54736314
Brown
Chrome Azurol S, Dye content: approx. 65%
C23H13Cl2Na3O9S
Mordant Blue 29, C.I. 43825
CC1=CC(=CC(=C1[O-])C(=O)[O-])C(=C2C=C(C(=O)C(=C2)C(=O)O)C)C3=C(C=CC(=C3Cl)S(=O)(=O)[O-])Cl.[Na+].[Na+].[Na+]
605.277
605.29
Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 216-787-0

Recommended Storage : Store at Room Temperature (15-30°C).

Product Identifier
  • Chrome Azurol S

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Chrome Azurol S

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.