Learn More
Corning™ Storage Mat III
Fits Corning™ 96 round well plates and blocks. Corning™ Storage Mat III can be applied manually or with a storage mat applicator.
Brand: Corning™ 3080
295.03 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
(This is the suggested replacement for 3092 and 3094)
- Fits Corning 96 round well plates and blocks
- Manufactured from EVA (ethyl vinyl acetate) and polymer is compatible with DMSO
- DNase- and RNase-free
- Can be applied manually or with our Storage Mat Applicator
- Nonsterile
Specifications
DNase and RNase-free | |
Non-sterile | |
25/Pk. | |
Round |
EVA, Polymer | |
100/Cs. | |
Storage Mat III | |
96-well Plate |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now