missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin D1 Antibody (CL12323), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21213-25ul
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
Cyclin D1 Monoclonal antibody specifically detects Cyclin D1 in Human samples. It is validated for Immunofluorescence
Specifica
Cyclin D1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
CL12323 | |
Immunocytochemistry/Immunofluorescence 2-10 ug/ml | |
B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31 | |
This antibody was developed against a Synthetic Peptide of P24385. Actual immunogen sequence is proprietary. | |
25 μg | |
Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers, Wnt Signaling Pathway | |
595 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Cyclin D1 Antibody (CL12323), Novus Biologicals™ > 25 μg; Unconjugated
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto