missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin D1 Antibody (CL12323), Novus Biologicals™
Mouse Monoclonal Antibody
378.00 EUR - 618.00 EUR
Specifications
Antigen | Cyclin D1 |
---|---|
Clone | CL12323 |
Dilution | Immunocytochemistry/Immunofluorescence 2-10 ug/ml |
Applications | Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18669154
|
Novus Biologicals
NBP3-21213-100ul |
100 μg |
618.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18631964
|
Novus Biologicals
NBP3-21213-25ul |
25 μg |
378.00 EUR
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin D1 Monoclonal antibody specifically detects Cyclin D1 in Human samples. It is validated for ImmunofluorescenceSpecifications
Cyclin D1 | |
Immunocytochemistry/Immunofluorescence 2-10 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31 | |
This antibody was developed against a Synthetic Peptide of P24385. Actual immunogen sequence is proprietary. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CL12323 | |
Immunofluorescence | |
Unconjugated | |
Mouse | |
Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers, Wnt Signaling Pathway | |
PBS, pH 7.2, 40% glycerol | |
595 | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cyclin D1 Antibody (CL12323), Novus Biologicals™