missing translation for 'onlineSavingsMsg'
Learn More

Cytosine-β-D-arabinofuranoside hydrochloride, MP BIOMEDICALS

Cytosine-β-D-Arabinofuranoside Hydrochloride

62.15 EUR - 208.00 EUR

Specifications

Product Type Nucleoside
Color White
Molecular Formula C9H14ClN3O5
CAS 69-74-9
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11398359
View Documents
MP Biomedicals
02100071.1
100 mg
62.15 EUR
100mg
Please to purchase this item. Need a web account? Register with us today!
11308369
View Documents
MP Biomedicals
02100071.5
500 mg
208.00 EUR
500mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Application Notes:

  • It is used as an antineoplastic and antiviral agent.

Usage Statement:

  • Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Key Applications:

  • Selective inhibitor of DNA synthesis
Specifications

Specifications

Nucleoside
White
C9H14ClN3O5
69-74-9
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytosine-β-D-arabinofuranoside hydrochloride, MP BIOMEDICALS

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.