Learn More
Fisherbrand™ Dedicated Platform
Fisherbrand™ Dedicated Platform is designed for use with a shaker.
395.00 EUR - 636.00 EUR
Specifications
Product Type | Dedicated Platform |
---|---|
Material | Aluminum, Stainless Steel |
For Use With (Equipment) | Mini Shaking Incubator |
Product Code | Brand | Holds | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | ||||||
16363246
|
Fisherbrand™
H1000-P-500 |
5 x 500 mL Flasks |
395.00 EUR
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
||||||
16353246
|
Fisherbrand™
H1000-P-250 |
8 x 250 mL mL Flasks |
395.00 EUR
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
||||||
16343246
|
Fisherbrand™
H1000-P-125 |
12 x 125 mL Flasks |
395.00 EUR
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
||||||
16373246
|
Fisherbrand™
H1000-P-MP |
4 Microplates |
636.00 EUR
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
||||||
Description
- Non-slip
- Rubber-coated surface
- For use with shaker
Specifications
Dedicated Platform | |
Aluminum, Stainless Steel | |
Mini Shaking Incubator |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Dedicated Platform