Learn More
Trajan Scientific and Medical™ Diluter/Dispenser Syringes for Hamilton Microlab 500™
Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for Hamilton Microlab 500™ has the knowledge and expertise that goes into manufacturing an SGE syringe.
Brand:  Trajan Scientific and Medical™ 004232
 View more versions of this product
Product Code. 15268473
  • 87.85 EUR / Each
Explore more special offers
Description
Description
Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for Hamilton Microlab 500™ has the knowledge and expertise that goes into manufacturing an SGE syringe.
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Diluter/Dispenser Syringe for Hamilton Microlab 500
Specifications
Specifications
60mm stroke, 6-32 UNC thread in plunger button
60 mm
50 μL
Plunger stop
Hamilton MicroLab 500™ Instrument Syringes
Luer Lock
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Trajan Scientific and Medical™ Diluter/Dispenser Syringes for Hamilton Microlab 500™ > Volume: 50μL; With plunger stop