missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase epsilon catalytic subunit A Antibody (3C5.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
Brand: Novus Biologicals NB100-115
This item is not returnable.
View return policy
Description
DNA Polymerase epsilon catalytic subunit A Monoclonal specifically detects DNA Polymerase epsilon catalytic subunit A in Human, Mouse, Mustelid, Primate, Drosophila (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
DNA Polymerase epsilon catalytic subunit A | |
Monoclonal | |
16 mg/mL | |
Western Blot 5 ug/ml, Immunohistochemistry reported in scientific literature (PMID 29935943), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:200 | |
DKFZp434F222, DNA polymerase epsilon catalytic subunit A, DNA polymerase II subunit A, EC 2.7.7, EC 2.7.7.7, POLE1FLJ21434, polymerase (DNA directed), epsilon | |
Mouse | |
220 kDa | |
0.1 mg | |
Primary | |
This is specific to the human DNA pol epsilon and does not recognize DNA pol alpha, beta or delta. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
3C5.1 | |
Unconjugated | |
Q9NRF9 | |
POLE | |
Human DNA Polymerase epsilon purified from HeLa cells. | |
Unpurified | |
RUO | |
5426 | |
Human, Mouse, Mustelid, Primate, Drosophila (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DNA Polymerase epsilon catalytic subunit A Antibody (3C5.1) - BSA Free, Novus Biologicals™ > 0.1 mg, Unconjugated
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction