missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase epsilon catalytic subunit A Antibody (3C5.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
219.00 EUR - 499.00 EUR
Specifications
Antigen | DNA Polymerase epsilon catalytic subunit A |
---|---|
Clone | 3C5.1 |
Concentration | 16 mg/mL |
Dilution | Western Blot 5 ug/ml, Immunohistochemistry reported in scientific literature (PMID 29935943), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18023975
|
Bio-Techne
NB100-115SS |
0.025 mg |
219.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18718583
|
Novus Biologicals
NB100-115 |
0.1 mg |
499.00 EUR
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNA Polymerase epsilon catalytic subunit A Monoclonal specifically detects DNA Polymerase epsilon catalytic subunit A in Human, Mouse, Mustelid, Primate, Drosophila (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
DNA Polymerase epsilon catalytic subunit A | |
16 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human, Mouse, Mustelid, Primate, Drosophila (Negative) | |
DKFZp434F222, DNA polymerase epsilon catalytic subunit A, DNA polymerase II subunit A, EC 2.7.7, EC 2.7.7.7, POLE1FLJ21434, polymerase (DNA directed), epsilon | |
POLE | |
IgG2a | |
Unpurified | |
This is specific to the human DNA pol epsilon and does not recognize DNA pol alpha, beta or delta. |
3C5.1 | |
Western Blot 5 ug/ml, Immunohistochemistry reported in scientific literature (PMID 29935943), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:200 | |
Monoclonal | |
Purified | |
RUO | |
Q9NRF9 | |
5426 | |
Human DNA Polymerase epsilon purified from HeLa cells. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
220 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DNA Polymerase epsilon catalytic subunit A Antibody (3C5.1) - BSA Free, Novus Biologicals™