Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, pointed, diamond-ground
Hollow glass stopper for sealing with draining tip.
Brand:  DWK Life Sciences
 View more versions of this product
Product Code. 10621872
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
DURAN™ hexagonal hollow plugs are used for sealing laboratory equipment such as volumetric flasks, mixing cylinders, ground joint flasks, etc. DURAN™ hexagonal hollow plugs are available in 2 standard versions - pointed - flat - Sizes analogous to DURAN™ conical ground joints. Cut quality: DG = Diamond Ground
  • Pointed
  • Diamond-ground
  • With a dripping syringe, for example, a condensate can be collected and the reaction medium recirculated.
  • hollow glass stopper
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Closure of vessels with standard ground joints
NS 12/21
Boro 3.3
100/Pk.
Autoclavable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences DURAN™ Hexagonal Stopper, pointed, diamond-ground > NS 12/21