Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, pointed, diamond-ground
Hollow glass stopper for sealing with draining tip.
1.09 EUR - 320.00 EUR
Specifications
Autoclavable Autoclavable
For Use With (Application) Closure of vessels with standard ground joints
View More Specs
Products 9
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
11772185
View Documents
DWK Life Sciences
NS 24/29
258.00 EUR
Pack of 100
Estimated Shipment: 27-03-2025
to see stock.
Add to basket
15864548
View Documents
DWK Life Sciences
286115406
NS 14/23
174.00 EUR
Pack of 100
In Stock
to see stock.
Edge
Add to basket
15911521
View Documents Promotion Details
DWK Life Sciences
NS 29/32
320.00 EUR
Pack of 100
In Stock
to see stock.
Add to basket
15921521
View Documents
DWK Life Sciences
286117904
NS 34/35
91.65 EUR
Pack of 10
Estimated Shipment: 10-04-2025
to see stock.
Add to basket
15884548
View Documents
DWK Life Sciences
NS 45/40
132.00 EUR
Pack of 10
In Stock
to see stock.
Add to basket
10621872
View Documents
DWK Life Sciences
NS 12/21 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10365191
View Documents
DWK Life Sciences
NS 10/19 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15874548
View Documents
DWK Life Sciences
NS 19/26
228.00 EUR
Pack of 100
In Stock
to see stock.
Add to basket
15854548
View Documents
DWK Life Sciences
NS 7/16
270.00 EUR
Pack of 100
In Stock
to see stock.
Add to basket
Description
Description
DURAN™ hexagonal hollow plugs are used for sealing laboratory equipment such as volumetric flasks, mixing cylinders, ground joint flasks, etc. DURAN™ hexagonal hollow plugs are available in 2 standard versions - pointed - flat - Sizes analogous to DURAN™ conical ground joints. Cut quality: DG = Diamond Ground
  • Pointed
  • Diamond-ground
  • With a dripping syringe, for example, a condensate can be collected and the reaction medium recirculated.
  • hollow glass stopper
Specifications
Specifications
Autoclavable
Closure of vessels with standard ground joints
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
DWK Life Sciences DURAN™ Hexagonal Stopper, pointed, diamond-ground