Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, pointed, diamond-ground
Hollow glass stopper for sealing with draining tip.
Brand:  DWK Life Sciences 286117904
 View more versions of this product
Product Code. 15921521
  • 91.65 EUR / Pack of 10
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Description
DURAN™ hexagonal hollow plugs are used for sealing laboratory equipment such as volumetric flasks, mixing cylinders, ground joint flasks, etc. DURAN™ hexagonal hollow plugs are available in 2 standard versions - pointed - flat - Sizes analogous to DURAN™ conical ground joints. Cut quality: DG = Diamond Ground
  • Pointed
  • Diamond-ground
  • With a dripping syringe, for example, a condensate can be collected and the reaction medium recirculated.
  • hollow glass stopper
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Closure of vessels with standard ground joints
NS 34/35
Borosilicate Glass
10/Pk.
Autoclavable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DWK Life Sciences DURAN™ Hexagonal Stopper, pointed, diamond-ground > NS 34/35