Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, pointed, diamond-ground
Hollow glass stopper for sealing with draining tip.
Brand:  DWK Life Sciences
 View more versions of this product
Product Code. 15854548
  • 270.00 EUR / Pack of 100
In Stock
to see stock.
Explore more special offers
Description
DURAN™ hexagonal hollow plugs are used for sealing laboratory equipment such as volumetric flasks, mixing cylinders, ground joint flasks, etc. DURAN™ hexagonal hollow plugs are available in 2 standard versions - pointed - flat - Sizes analogous to DURAN™ conical ground joints. Cut quality: DG = Diamond Ground
  • Pointed
  • Diamond-ground
  • With a dripping syringe, for example, a condensate can be collected and the reaction medium recirculated.
  • hollow glass stopper
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Closure of vessels with standard ground joints
NS 7/16
Borosilicate Glass
100/Pk.
Autoclavable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences DURAN™ Hexagonal Stopper, pointed, diamond-ground > NS 7/16