Learn More
Eurofins DiscoverX DUSP22 Protein
N-terminal GST-tagged, recombinant, full length, human DUSP22, for use in Phosphatase Assays.
Brand: Eurofins DiscoverX 14-641M

Description
DUSP22 activates the Jnk signaling pathway. It dephosphorylates and deactivates p38 and stress-activated protein kinase/c-Jun N-terminal kinase (SAPK/JNK) (By similarity)
GenBank NM_020185.
Specifications
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
250 μg | |
Recombinant | |
Recombinant enzyme expressed in E. coli cells | |
≥70% |
47.3 kDa | |
Human | |
N-terminal GST | |
Full-length | |
DUSP22, active |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.