Learn More
Thermo Scientific™ Crimper and Decrimper Jaw Sets for Programmable Electronic High-Power Crimp Stations
Optimize your Thermo Scientific™ Programmable Electronic High-Power Crimp Stations with these crimper and decrimper sets.
Brand:  Thermo Scientific™ ECMJ-11
338.16 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15257308
  • 339.85 EUR / Each
Description
Optimize your Thermo Scientific™ Programmable Electronic High-Power Crimp Station with these crimper and decrimper sets.
Available in a variety of sizes and formats.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Ea.
Programmable Electronic High-Power Crimp Station, 11 mm Vial Closure
Industrial Chromatography
Electronic Crimper
11 mm
Documents
One moment while we fetch your results.
Product Title
Thermo Scientific™ Crimper and Decrimper Jaw Sets for Programmable Electronic High-Power Crimp Stations >