Learn More
Gilson™ Extraction Cartridge Cap
Gilson offers a variety of vials and caps in bulk for a number of applications.
Brand:  Gilson™ 2954699
593.21 EUR valid until 2025-05-31
Use promo code "24127" to get your promotional price.
 View more versions of this product
Product Code. 16666155
  • 646.00 EUR / Pack of 1000
In Stock
Description
  • ASPEC Caps
  • Gilson offers a variety of vials and caps in bulk for a number of applications.
Specifications
Specifications
ASPEC Caps
3 mL
Natural PE
1000 Pk.
Product Title
Gilson™ Extraction Cartridge Cap > 3 mL