Learn More
Fisherbrand™ Glass Pear-Shaped Separating Funnel with Glass Stopcock
Separates mixtures into immiscible solvents. Fisherbrand™ Glass Pear-Shaped Separating Funnel with Glass Stopcock is available with interchangeable glass or PTFE stopcock, but supplied with a glass and plastic stopper.
Brand:  Fisherbrand™ FB51753
 View more versions of this product
Product Code. 11922138
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Separates mixtures into immiscible solvents. Fisherbrand™ Glass Pear-Shaped Separating Funnel with Glass Stopcock is available with interchangeable glass or PTFE stopcock, but supplied with a glass and plastic stopper.
  • Easy-open stopper helps to reduce pressure buildup during processing.
  • Constructed in chemical resistant borosilicate glass.
  • ISO 4800 Certified.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
500 mL
ISO 4800
Glass
24/29
Borosilicate Glass
1PK
Glass, Plastic
Separating funnel
SDS
Documents Certificates
Product Title
Fisherbrand™ Glass Pear-Shaped Separating Funnel with Glass Stopcock > Capacity: 500mL