Learn More
Heidolph™ Instruments Hei-VAP Automatic Evaporation plus Package, EU-plug
The Hei-VAP Automatic Evaporation plus Package features a motor lift, Woulff bottle, condenser, vacuum pump, and tubing set.
Brand:  Heidolph™ Instruments 574-38310-00
10599.78 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Product Code. 17249039
  • 12060.00 EUR / Each
Estimated Shipment: 13-06-2025
Description
  • Hei-VAP Expert Control with motor lift and coated vertical glassware set G3B
  • Vacuum pump Rotavac Vario Control
  • Condenser for Rotavac Vario
  • Tubing set
  • Woulff bottle
  • AUTOaccurate-sensor
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Motor Lift Rotary Evaporator
EU-plug
Product Title
Heidolph™ Instruments Hei-VAP Automatic Evaporation plus Package, EU-plug > Type: Motor Lift Rotary Evaporator