Learn More
Histone H3.1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-81964
396.80 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
 View more versions of this product
Product Code. 18746503
  • 496.00 EUR / 0.10mL
Explore more special offers
Description
Description
Histone H3.1 Polyclonal specifically detects Histone H3.1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:200
HIST1H3E
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Histone H3.1 Antibody, Novus Biologicals™ > 0.1 mg; Unconjugated