missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3.1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
197.00 EUR - 496.00 EUR
Specifications
Antigen | Histone H3.1 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:200 |
Applications | Immunofluorescence, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18245544
|
Bio-Techne
NB100-81964-0.025ML |
0.025 mL |
197.00 EUR
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18746503
|
Novus Biologicals
NB100-81964 |
496.00 EUR
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Histone H3.1 Polyclonal specifically detects Histone H3.1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Histone H3.1 | |
Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:200 | |
Polyclonal | |
Rabbit | |
Epigenetics, Histones and Modified Histones | |
P68431 | |
8353 | |
IgG | |
Affinity Purified | |
17 kDa |
1.0 mg/mL | |
Immunofluorescence, Western Blot, Immunocytochemistry | |
Unconjugated | |
RUO | |
Rat, Human, Mouse | |
H3 histone family, member D, H3.1HIST1H3D, H3/d, H3FA, H3FB, H3FC, H3FDHIST1H3F, H3FF, H3FH, H3FI, H3FJ, H3FK, H3FL, HIST1H3A, HIST1H3B, HIST1H3C, HIST1H3G, HIST1H3H, HIST1H3I, HIST1H3J, histone 1, H3e, histone cluster 1, H3e, histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l | |
HIST1H3E | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Histone H3.1 Antibody, Novus Biologicals™