Learn More
Histone H3.1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-81964
396.80 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
Histone H3.1 Polyclonal specifically detects Histone H3.1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:200 | |
HIST1H3E |
Product Content Correction
Product Title
Histone H3.1 Antibody, Novus Biologicals™ > 0.1 mg; Unconjugated
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction