Learn More
R&D Systems™ HIV-1 Gag p24 DuoSet ELISA
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand: R&D Systems™ DY7360-05
280.80 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
The HIV-1 Gag p24 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies viral HIV-1 Gag p24.
Specifications
15.60 - 1000 pg/mL | |
ELISA | |
Viral | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Solid Phase Sandwich ELISA | |
155030 | |
1 Kit (5 Plates) | |
HIV-1 Gag p24 |
Product Content Correction
Product Title
R&D Systems™ HIV-1 Gag p24 DuoSet ELISA > Quantity: 1 Kit (for 5 Plates); Host Species: Viral
Spot an opportunity for improvement?Share a Content Correction