missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP L Antibody (4D11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NB120-6106SS
165.60 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
hnRNP L Monoclonal specifically detects hnRNP L in Human, Mouse, Rat, Amphibian, Bovine, Chicken, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
hnRNP L | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:5000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:2000 | |
FLJ35509, heterogeneous nuclear ribonucleoprotein L, hnRNP L, hnRNP-L, HNRPL | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Monkey, Chicken and Newt. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
4D11 | |
Unconjugated | |
P14866, Q8R081 | |
HNRNPL | |
Human hnRNP proteins from HeLa cells purified by affinity chromatography on ssDNA agarose. | |
0.025 mL | |
Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers | |
3191 | |
Human, Mouse, Rat, Amphibian, Bovine, Chicken, Primate | |
Purified |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
hnRNP L Antibody (4D11) - BSA Free, Novus Biologicals™ > 0.025mL; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction