missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP L Antibody (4D11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
207.00 EUR - 449.00 EUR
Specifications
Antigen | hnRNP L |
---|---|
Clone | 4D11 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:5000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:2000 |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18246960
|
Bio-Techne
NB120-6106SS |
0.025 mL |
207.00 EUR
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18770524
|
Novus Biologicals
NB120-6106 |
0.1 mL |
449.00 EUR
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
hnRNP L Monoclonal specifically detects hnRNP L in Human, Mouse, Rat, Amphibian, Bovine, Chicken, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
hnRNP L | |
1.0 mg/mL | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers | |
P14866, Q8R081 | |
3191 | |
Human hnRNP proteins from HeLa cells purified by affinity chromatography on ssDNA agarose. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
4D11 | |
Western Blot 1:5000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Amphibian, Bovine, Chicken, Primate | |
FLJ35509, heterogeneous nuclear ribonucleoprotein L, hnRNP L, hnRNP-L, HNRPL | |
HNRNPL | |
IgG1 | |
Protein G purified | |
Human, Mouse, Rat, Bovine, Monkey, Chicken and Newt. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
hnRNP L Antibody (4D11) - BSA Free, Novus Biologicals™