missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP40 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA575171
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Heat shock proteins (HSP) are expressed in response to various biological stresses, including high temperatures. There are several major families of HSPs including HSP70, HSP90 and HSP100. HSP40 is a new member of the hsp40 family, exhibits similar expression profile to that of hsc70 in mammalian cells.
Specifications
HSP40 | |
Polyclonal | |
Unconjugated | |
DNAJB1 | |
DnaJ homolog subfamily B member 1; DnaJ protein homolog 1; Heat shock 40 kDa protein 1; HSP 40; Heat shock protein 40; Human DnaJ protein 1; hDj-1; DNAJB1; DNAJ1; HDJ1; HSPF1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3337, 361384, 81489 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P25685, Q9QYJ3 | |
DNAJB1 | |
Synthetic peptide corresponding to amino acids 270-320 of Human HSP40. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HSP40 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction