Learn More
Human CD42b/GPIb alpha APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB4067A
406.40 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
Description
CD42b/GPIb alpha Monoclonal specifically detects CD42b/GPIb alpha in Human samples. It is validated for Flow Cytometry.
Specifications
CD42b/GPIb alpha | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Antigen CD42b-alpha, BP1BA, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP1BA, GPIb alpha, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2811 | |
Human | |
Purified |
Flow Cytometry | |
486805 | |
Flow Cytometry 10 uL/10^6 cells | |
P07359.1 | |
GP1BA | |
Mouse myeloma cell line NS0-derived recombinant human CD42b/GPIb alpha His17-Leu505 Accession # P07359.1 | |
100 Tests | |
Primary | |
Detects human CD42b/GPIb alpha in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |